(3-Brom-4-methoxyphenyl)acetonitril ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-(3-Bromo-4-methoxyphenyl)acetonitrile
L09840.09
L09840.04
L09840.09EA
122
EUR
L09840.09
L09840.04
(3-Brom-4-methoxyphenyl)acetonitril ≥99%
(3-Brom-4-methoxyphenyl)acetonitril
Purity: 99%
CAS No.: 772-59-8
Molecular Formula: C9H8BrNO
Formula Weight: 226.08
Melting Point: 50-53°
Boiling Point: 175-178°/6mm
UN No.: 3439
MDL No.: MFCD00016391
Formel:
C₉H₈BrNO Molecular Weight: 226,08 g/mol Siedepunkt: 175…178 °C (6 mmHg) Schmelzpunkt: 50…53 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016391 CAS-Nummer: 772-59-8 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...