(2-Brom-4,5-dimethoxyphenyl)acetonitril
Lieferant: FLUOROCHEM
Synonyme:
2-(2-Bromo-4,5-dimethoxyphenyl)acetonitrile
018271-1G
018271-5G
FLUO018271-1GEA
39.9
EUR
FLUO018271-1G
FLUO018271-5G
(2-Brom-4,5-dimethoxyphenyl)acetonitril
(2-Brom-4,5-dimethoxyphenyl)acetonitril
Formel:
C₁₀H₁₀BrNO₂ Molecular Weight: 256.1000 g/mol |
MDL:
MFCD00016385 CAS-Nummer: 51655-39-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...