3-Chlor-4,5-diaminobenzotrifluorid
Lieferant: Thermo Scientific
Synonyme:
3-Chloro-5-trifluoromethyl-o-phenylenediamine
BTB05127.1
BTB05127EA
MAYBBTB05127.1EA
0
EUR
MAYBBTB05127.1
MAYBBTB05127EA
3-Chlor-4,5-diaminobenzotrifluorid
3-Chlor-4,5-diaminobenzotrifluorid
Formel:
C₇H₆ClF₃N₂ Molecular Weight: 210,59 g/mol |
MDL:
MFCD00042198 CAS-Nummer: 132915-80-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...