1,1,1-Trifluor-3-(phenethylamino)-2-propanol

Lieferant: BIONET RESEARCH

Synonyme: 3-Phenethylamino-1,1,1-trifluoropropan-2-ol

7P-037.500 7P-037.1 7P-037.5 7P-037.10
BIOT7P-037.500EA 0 EUR
BIOT7P-037.500 BIOT7P-037.1 BIOT7P-037.5 BIOT7P-037.10
1,1,1-Trifluor-3-(phenethylamino)-2-propanol
1,1,1-Trifluor-3-(phenethylamino)-2-propanol
Formel: C₁₁H₁₄F₃NO
MDL: MFCD03001278
CAS-Nummer: 400878-20-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR