Diethyl(4-chlorbenzyl)phosphonat ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Diethyl 4-chlorobenzylphosphonate
L00965.09
L00965.18
L00965.09EA
53.5
EUR
L00965.09
L00965.18
Diethyl(4-chlorbenzyl)phosphonat ≥98%
Diethyl(4-chlorbenzyl)phosphonat
Purity: 98+%
CAS No.: 39225-17-7
Molecular Formula: C11H16ClO3P
Formula Weight: 262.67
Flash Point:
Formel:
C₁₁H₁₆ClO₃P Molecular Weight: 262.67 g/mol Siedepunkt: 153…155 °C (0.6 mmHg) Dichte: 1.082 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051568 CAS-Nummer: 39225-17-7 EINECS: 254-365-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...