(4-Aminobenzyl)phosphonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (4-Aminobenzyl)phosphonic Acid

A5898-250MG A5898-1G
SIALA5898-250MGEA 83.2 EUR
SIALA5898-250MG SIALA5898-1G
(4-Aminobenzyl)phosphonsäure, Sigma-Aldrich®
(4-Aminobenzyl)phosphonsäure
Formel: C₇H₁₀NO₃P
Molecular Weight: 187,13 g/mol
MDL: MFCD00070204
CAS-Nummer: 5424-27-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR