5-Brom-8-fluorchinolin

Lieferant: COMBI-BLOCKS

HC-2356-5G HC-2356-1G
COBBHC-2356-5GEA 798 EUR
COBBHC-2356-5G COBBHC-2356-1G
5-Brom-8-fluorchinolin
5-Bromo-8-fluoroquinoline
Formel: C₉H₅BrFN
Molecular Weight: 226,0500 g/mol
MDL: MFCD11855895
CAS-Nummer: 1133115-78-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR