6-Brom-5,8-difluorchinolin

Lieferant: COMBI-BLOCKS

HC-2331-5G HC-2331-1G
COBBHC-2331-5GEA 451 EUR
COBBHC-2331-5G COBBHC-2331-1G
6-Brom-5,8-difluorchinolin
6-Bromo-5,8-difluoroquinoline
Formel: C₉H₄BrF₂N
MDL: MFCD11855886
CAS-Nummer: 1133115-72-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR