8-Brom-5-fluorchinolin

Lieferant: COMBI-BLOCKS

HC-2022-5G HC-2022-1G
COBBHC-2022-5GEA 1010 EUR
COBBHC-2022-5G COBBHC-2022-1G
8-Brom-5-fluorchinolin
8-Bromo-5-fluoroquinoline
Formel: C₉H₅BrFN
Molecular Weight: 226,0451 g/mol
MDL: MFCD09907839
CAS-Nummer: 917251-99-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR