4-Chlor-6-fluorchinolin
Lieferant: FLUOROCHEM
CFF952.1
005481-1G
005481-500MG
CHMPCFF952.1EA
522
EUR
CHMPCFF952.1
FLUO005481-1G
FLUO005481-500MG
4-Chlor-6-fluorchinolin
4-Chloro-6-fluoroquinoline
Formel:
C₉H₅ClFN Molecular Weight: 181.60 g/mol |
MDL:
MFCD00278783 CAS-Nummer: 391-77-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...