4-Chlor-8-fluorchinolin
Lieferant: Thermo Scientific
|
Achtung
|
H50541.03
H50541.06
H50541.03EA
117
EUR
H50541.03
H50541.06
4-Chlor-8-fluorchinolin
4-Chloro-8-fluoroquinoline
CAS No.: 63010-72-0
Molecular Formula: C9H5ClFN
Formula Weight: 181.60
MDL No.: MFCD00278785
Formel:
C₉H₅ClFN Molecular Weight: 181,60 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00278785 CAS-Nummer: 63010-72-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...