2-Fluorchinolin-3-boronsäure

Lieferant: COMBI-BLOCKS

BB-8481-1G BB-8481-250M
COBBBB-8481-1GEA 474 EUR
COBBBB-8481-1G COBBBB-8481-250M
2-Fluorchinolin-3-boronsäure
2-Fluorchinolin-3-boronsäure
Formel: C₉H₇BFNO₂
Molecular Weight: 190,97 g/mol
MDL: MFCD05664131
CAS-Nummer: 745784-10-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR