5-Fluorchinolin-8-boronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(5-Fluoroquinolin-8-yl)boronic acid
BB-5327-1G
BB-5327-250M
COBBBB-5327-1GEA
864
EUR
COBBBB-5327-1G
COBBBB-5327-250M
5-Fluorchinolin-8-boronsäure
5-Fluorchinolin-8-boronsäure
Formel:
C₉H₇BFNO₂ Molecular Weight: 190,9668 g/mol |
MDL:
MFCD11504850 CAS-Nummer: 1072951-45-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...