4-Brom-6-chlor-2-(trifluormethyl)chinolin

Lieferant: COMBI-BLOCKS

HC-2149-5G HC-2149-1G
COBBHC-2149-5GEA 595 EUR
COBBHC-2149-5G COBBHC-2149-1G
4-Brom-6-chlor-2-(trifluormethyl)chinolin
4-Bromo-6-chloro-2-(trifluoromethyl)quinoline
Formel: C₁₀H₄BrClF₃N
MDL: MFCD11504906
CAS-Nummer: 18706-32-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR