4-Brom-6-fluor-2-(trifluormethyl)chinolin

Lieferant: COMBI-BLOCKS

HC-2171-5G HC-2171-1G
COBBHC-2171-5GEA 595 EUR
COBBHC-2171-5G COBBHC-2171-1G
4-Brom-6-fluor-2-(trifluormethyl)chinolin
4-Bromo-6-fluoro-2-(trifluoromethyl)quinoline
Formel: C₁₀H₄BrF₄N
MDL: MFCD11504912
CAS-Nummer: 31009-33-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR