4,7-Dichlor-8-methyl-2-(trifluormethyl)chinolin

Lieferant: COMBI-BLOCKS

HC-2172-1G HC-2172-5G
COBBHC-2172-1GEA 149 EUR
COBBHC-2172-1G COBBHC-2172-5G
4,7-Dichlor-8-methyl-2-(trifluormethyl)chinolin
4,7-Dichloro-8-methyl-2-(trifluoromethyl)quinoline
Formel: C₁₁H₆Cl₂F₃N
MDL: MFCD08448261
CAS-Nummer: 887350-78-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR