4-Chlor-6-methyl-2-(trifluormethyl)chinolin

Lieferant: FLUOROCHEM

004505-1G 004505-500MG
FLUO004505-1GEA 159 EUR
FLUO004505-1G FLUO004505-500MG
4-Chlor-6-methyl-2-(trifluormethyl)chinolin
4-Chloro-6-methyl-2-(trifluoromethyl)quinoline
Formel: C₁₁H₇ClF₃N
Molecular Weight: 245.630 g/mol
MDL: MFCD00153097
CAS-Nummer: 1701-26-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR