N-Methyldicyclohexylamin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
N,N-Dicyclohexylmethylamine, N-Cyclohexyl-N-methylcyclohexanamine
M-Clarity™ Qualitätsstufe = MQ200
294942-250G
294942-1KG
SIAL294942-250GEA
98.7
EUR
SIAL294942-250G
SIAL294942-1KG
N-Methyldicyclohexylamin, Sigma-Aldrich®
N-Methyldicyclohexylamin
Formel:
C₁₃H₂₅N Molecular Weight: 195.3443 g/mol |
MDL:
MFCD00014289 CAS-Nummer: 7560-83-0 EINECS: 231-453-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...