Diphenylphosphonat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIALD210803-100GEA 54.1 EUR
SIALD210803-100G SIALD210803-500G
Diphenylphosphonat, Sigma-Aldrich®
Diphenylphosphonat
Formel: C₁₂H₁₁O₃P
Molecular Weight: 234,19 g/mol
MDL: MFCD00044497
CAS-Nummer: 4712-55-4
EINECS: 225-202-8
UN: 3265
ADR: 8,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR