Chlortitantriisopropoxid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
250627-100G
250627-800G
SIAL250627-100GEA
110
EUR
SIAL250627-100G
SIAL250627-800G
Chlortitantriisopropoxid, Sigma-Aldrich®
Chlortitantriisopropoxid
Formel:
C₉H₂₁ClO₃Ti Molecular Weight: 260,58 g/mol |
MDL:
MFCD00009861 CAS-Nummer: 20717-86-6 UN: 2921 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...