Methyl-4-brom-1-naphthoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 4-bromonaphthalene-1-carboxylate

CA-4365-5G CA-4365-1G
COBBCA-4365-5GEA 190 EUR
COBBCA-4365-5G COBBCA-4365-1G
Methyl-4-brom-1-naphthoat
Methyl-4-brom-1-naphthoat
Formel: C₁₂H₉BrO₂
Molecular Weight: 265,1027 g/mol
MDL: MFCD09475914
CAS-Nummer: 35615-97-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR