Methyl-6-brom-2-naphthoat ≥98%
Lieferant: Thermo Scientific
Synonyme:
Methyl 6-bromo-2-naphthoate
B25106.06
B25106.14
B25106.06EA
58.8
EUR
B25106.06
B25106.14
Methyl-6-brom-2-naphthoat ≥98%
Methyl-6-brom-2-naphthoat
Purity: 98%
CAS No.: 33626-98-1
Molecular Formula: C12H9BrO2
Formula Weight: 265.11
Melting Point: 121-123°
Beilstein Reference: 2578943
Formel:
C₁₂H₉BrO₂ Molecular Weight: 265,11 g/mol Schmelzpunkt: 121…123 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00100408 CAS-Nummer: 33626-98-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...