Methyl-6-brom-2-naphthoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 6-bromo-2-naphthoate

CA-4585-25G CA-4585-5G CA-4585-1G CA-4585-100G
COBBCA-4585-25GEA 181 EUR
COBBCA-4585-25G COBBCA-4585-5G COBBCA-4585-1G COBBCA-4585-100G
Methyl-6-brom-2-naphthoat
Methyl-6-brom-2-naphthoat
Formel: C₁₂H₉BrO₂
Molecular Weight: 265,11 g/mol
MDL: MFCD00100408
CAS-Nummer: 33626-98-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR