4-Brom-1-naphthoesäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Bromo-1-naphthalenecarboxylic acid

CA-4257-5G CA-4257-25G CA-4257-1G
COBBCA-4257-5GEA 0 EUR
COBBCA-4257-5G COBBCA-4257-25G COBBCA-4257-1G
4-Brom-1-naphthoesäure
4-Brom-1-naphthoesäure
Formel: C₁₁H₇BrO₂
MDL: MFCD00094221
CAS-Nummer: 16650-55-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR