6-Brom-2-naphthoesäure
Lieferant: COMBI-BLOCKS
OS-7426-5G
OS-7426-25G
OS-7426-1G
OS-7426-100G
COBBOS-7426-5GEA
0
EUR
COBBOS-7426-5G
COBBOS-7426-25G
COBBOS-7426-1G
COBBOS-7426-100G
6-Brom-2-naphthoesäure
6-Brom-2-naphthoesäure
Formel:
C₁₁H₇BrO₂ |
MDL:
MFCD01075720 CAS-Nummer: 5773-80-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...