8-Brom-1-naphthoesäure
Lieferant: Thermo Scientific
BTB12326.250
BTB12326DA
AB145134.0001
MAYBBTB12326.250EA
0
EUR
MAYBBTB12326.250
MAYBBTB12326DA
ABCRAB145134.0001
8-Brom-1-naphthoesäure
8-Brom-1-naphthoesäure
Formel:
C₁₁H₇BrO₂ |
MDL:
MFCD00029357 CAS-Nummer: 1729-99-3 EINECS: 217-045-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...