Methyl-2-naphthoat
Lieferant: CHEMPUR
Synonyme:
Methyl 2-naphthoate
FL13169.POR
013169-5G
013169-1G
013169-25G
CHMPFL13169.POREA
0
EUR
CHMPFL13169.POR
FLUO013169-5G
FLUO013169-1G
FLUO013169-25G
Methyl-2-naphthoat
Methyl-2-naphthoat
Formel:
C₁₂H₁₀O₂ Molecular Weight: 186,21 g/mol |
MDL:
MFCD00017345 CAS-Nummer: 2459-25-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...