1-Amino-4-naphthol Hydrochlorid

Lieferant: CHEMPUR

Synonyme: 4-Amino-1-naphthol hydrochloride

FL13411.POR 013411-5G
CHMPFL13411.POREA 0 EUR
CHMPFL13411.POR FLUO013411-5G
1-Amino-4-naphthol Hydrochlorid
1-Amino-4-naphthol Hydrochlorid
Formel: C₁₀H₁₀ClNO
Molecular Weight: 195,65 g/mol
MDL: MFCD00012562
CAS-Nummer: 5959-56-8
EINECS: 227-727-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR