1-(2-Naphthoyl)-3,3,3-trifluoraceton
Lieferant: Thermo Scientific
Synonyme:
4,4,4-Trifluoro-1-(naphthalen-2-yl)butane-1,3-dione, 4,4,4-Trifluoro-1-(2-naphthyl)-1,3-butanedione
SB01917.1
SB01917.50
MAYBSB01917.1EA
0
EUR
MAYBSB01917.1
MAYBSB01917.50
1-(2-Naphthoyl)-3,3,3-trifluoraceton
1-(2-Naphthoyl)-3,3,3-trifluoraceton
Formel:
C₁₄H₉F₃O₂ Molecular Weight: 266.22 g/mol |
MDL:
MFCD00054501 CAS-Nummer: 893-33-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...