5-Brom-2-methoxy-6-methylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromo-6-methoxy-2-picoline, 3-Bromo-6-methoxy-2-methylpyridine

PY-7908-5G PY-7908-1G
COBBPY-7908-5GEA 71.2 EUR
COBBPY-7908-5G COBBPY-7908-1G
5-Brom-2-methoxy-6-methylpyridin
5-Brom-2-methoxy-6-methylpyridin
Formel: C₇H₈BrNO
Molecular Weight: 202,05 g/mol
MDL: MFCD03094943
CAS-Nummer: 126717-59-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR