5-Brom-2-methoxy-4-methylpyridin
Lieferant: COMBI-BLOCKS
Synonyme:
5-Bromo-2-methoxy-4-methylpyridine
PY-7843-5G
PY-7843-1G
COBBPY-7843-5GEA
102
EUR
COBBPY-7843-5G
COBBPY-7843-1G
5-Brom-2-methoxy-4-methylpyridin
5-Brom-2-methoxy-4-methylpyridin
Formel:
C₇H₈BrNO Molecular Weight: 202.0485 g/mol |
MDL:
MFCD04039980 CAS-Nummer: 164513-39-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...