5-Brom-2-methoxy-3-methylpyridin ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
5-Bromo-2-methoxy-3-methylpyridine
H50113.03
H50113.06
H50113.03EA
158
EUR
H50113.03
H50113.06
5-Brom-2-methoxy-3-methylpyridin ≥97%
5-Brom-2-methoxy-3-methylpyridin
Purity: 97%
CAS No.: 760207-87-2
Molecular Formula: C7H8BrNO
Formula Weight: 202.05
Refractive Index: 1.5540
Formel:
C₇H₈BrNO Molecular Weight: 202.05 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07375124 CAS-Nummer: 760207-87-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...