Thiophen-3-acetonitril ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-(Thiophen-3-yl)acetonitrile, 3-Thiopheneacetonitrile

A12805.06 A12805.14
A12805.06EA 63.6 EUR
A12805.06 A12805.14
Thiophen-3-acetonitril ≥98%
Thiophen-3-acetonitril

Purity: 98% CAS No.: 13781-53-8 Molecular Formula: C6H5NS Formula Weight: 123.18 Flash Point: 119°(246°F) Refractive Index: 1.5430 Boiling Point: 240-242° Beilstein Reference: 107774 UN No.: 3276 MDL No.: MFCD00005471

Formel: C₆H₅NS
Molecular Weight: 123,18 g/mol
Siedepunkt: 240…242 °C
Dichte: 1,080
Flammpunkt: 119 °C (246 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005471
CAS-Nummer: 13781-53-8
EINECS: 237-433-1
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR