Benzo[b]thiophen-2-methanol

Lieferant: FLUOROCHEM

Synonyme: 2-(Hydroxymethyl)benzo[b]thiophene, Benzo[b]thiophen-2-ylmethanol

CFF484.2.5 CFF484.10 017493-2.5G 017493-10G
CHMPCFF484.2.5EA 375 EUR
CHMPCFF484.2.5 CHMPCFF484.10 FLUO017493-2.5G FLUO017493-10G
Benzo[b]thiophen-2-methanol
Benzo[b]thiophen-2-methanol
Formel: C₉H₈OS
Molecular Weight: 164,22 g/mol
MDL: MFCD02682004
CAS-Nummer: 17890-56-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR