Benzo[b]thiophen-3-methanol

Lieferant: Thermo Scientific

Synonyme: 3-(Hydroxymethyl)benzo[b]thiophene

CC12309.250 CC12309DA
MAYBCC12309.250EA 0 EUR
MAYBCC12309.250 MAYBCC12309DA
Benzo[b]thiophen-3-methanol
Benzo[b]thiophen-3-methanol
Formel: C₉H₈OS
CAS-Nummer: 5381-24-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR