(S)-(-)-1,2,3,4-Tetrahydro-1-naphthoesäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(S)-1,2,3,4-Tetrahydro-1-naphthoic acid, (1S)-1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid
H27612.MD
H27612.03
H27612.MDEA
29
EUR
H27612.MD
H27612.03
(S)-(-)-1,2,3,4-Tetrahydro-1-naphthoesäure ≥98%
(S)-(-)-1,2,3,4-Tetrahydro-1-naphthoesäure
Purity: 98%
CAS No.: 85977-52-2
Molecular Formula: C11H12O2
Formula Weight: 176.22
Melting Point: 55-57°
Optical Rotation: -12° (c=1 in ethanol)
Formel:
C₁₁H₁₂O₂ Molecular Weight: 176.22 g/mol Schmelzpunkt: 55…57 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368365 CAS-Nummer: 85977-52-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...