Menadionnatriumbisulfit ≥95% (durch TLC), Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
2-Methyl-1,4-naphthoquinone sodium bisulphite, Menadione bisulphite sodium
M-Clarity™ Qualitätsstufe = MQ200
M5750-100G
M5750-1KG
M5750-25G
SIALM5750-100GEA
125
EUR
SIALM5750-100G
SIALM5750-1KG
SIALM5750-25G
Menadionnatriumbisulfit ≥95% (durch TLC), Sigma-Aldrich®
Menadionnatriumbisulfit
Water soluble vitamin K3.
Menadione sodium bisulfite is a water-soluble form of menadione, which belongs to the Vitamin K class of compounds. These are necessary for the biosynthesis of prothrombin and other blood clotting factors. Menadione is a prothrombogenic compound and is used as a model quinone in cell culture and in vivo investigations.
Formel:
C₁₁H₉NaO₅S Molecular Weight: 276,24 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00051456 CAS-Nummer: 130-37-0 EINECS: 204-987-0 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...