5-Acetyl-2,2-dimethyl-1,3-dioxan-4,6-dion (5-Acetylmeldrumsäure)

Lieferant: BIONET RESEARCH

Synonyme: 5-Acetyl-2,2-dimethyl-1,3-dioxane-4,6-dione

1D-012.500 1D-012.1 1D-012.5 1D-012.10
BIOT1D-012.500EA 0 EUR
BIOT1D-012.500 BIOT1D-012.1 BIOT1D-012.5 BIOT1D-012.10
5-Acetyl-2,2-dimethyl-1,3-dioxan-4,6-dion (5-Acetylmeldrumsäure)
5-Acetyl-2,2-dimethyl-1,3-dioxan-4,6-dion (5-Acetylmeldrumsäure)
Formel: C₈H₁₀O₅
Molecular Weight: 186.1620 g/mol
MDL: MFCD03425724
CAS-Nummer: 72324-39-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR