Poly(2-ethyl-2-oxazolin), M.W. ~200,000
Lieferant: Thermo Fisher Scientific
42397.22
42397.36
42397.A4
42397.22EA
43.6
EUR
42397.22
42397.36
42397.A4
Poly(2-ethyl-2-oxazolin), M.W. ~200,000
Poly(2-ethyl-2-oxazolin)
Formel:
(C₅H₉NO)x Molecular Weight: 200000 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
25805-17-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...