Poly(2-ethyl-2-oxazolin), M.W. ~50,000

Lieferant: Thermo Scientific

42396.22 42396.36 42396.A4
42396.22EA 40.2 EUR
42396.22 42396.36 42396.A4
Poly(2-ethyl-2-oxazolin), M.W. ~50,000
Poly(2-ethyl-2-oxazolin)

CAS No.: 25805-17-8 Molecular Formula: [-N(COC2H5)CH2CH2-]n Physical Form: Solid MDL No.: MFCD00134377

Formel: (C₅H₉NO)x
Molecular Weight: 200000 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 25805-17-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR