Fmoc-Met-OH
Lieferant: BACHEM BIOCHEMICA
BACE4003154.0100EA
110
EUR
BACE4003154.0100
BACE4003154.0500
BACE4003154.0025
Fmoc-Met-OH
Fmoc-Met-OH
Formel:
C₂₀H₂₁NO₄S Molecular Weight: 371,46 g/mol |
MDL:
MFCD00037134 CAS-Nummer: 71989-28-1 EINECS: 276-258-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...