4-Hydroxy-2-chinolincarbonsäure Hydrat 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Hydroxyquinaldic acid hydrate, Kynurenic acid hydrate
121910010
121910050
121910250
ACRO121910010EA
33
EUR
ACRO121910010
ACRO121910050
ACRO121910250
4-Hydroxy-2-chinolincarbonsäure Hydrat 98%
4-Hydroxy-2-chinolincarbonsäure Hydrat
Formel:
C₁₀H₇NO₃·xH₂O Molecular Weight: 189,17 g/mol (anhy) Schmelzpunkt: 275 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006753 CAS-Nummer: 345909-35-5 EINECS: 207-751-5 Merck Index: 13,05342 |
Spezifikation Testergebnisse
Appearance | White to yellow to brown Powder |
Infrared spectrum | Conforms |
Water | 7 to 10 % (K.F.) |
Titration excess NaOH | 97.5 to 102.5 % (On dry substance) |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...