4-Hydroxy-2-chinolincarbonsäure Hydrat ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Hydroxyquinaldic acid hydrate, Kynurenic acid hydrate

A12602.06 A12602.14
A12602.06EA 93.8 EUR
A12602.06 A12602.14
4-Hydroxy-2-chinolincarbonsäure Hydrat ≥98%
4-Hydroxy-2-chinolincarbonsäure Hydrat

Broad spectrum excitatory amino acid antagonist.


4-Hydroxyquinoline-2-carboxylic acid hydrate is used as an intermediate in synthetic chemistry. It acts as an antiexcitotoxic and anticonvulsant. It finds application in certain neurobiological disorders therapy.

Formel: C₁₀H₇NO₃·xH₂O
Molecular Weight: 189,17 g/mol (anhy)
Schmelzpunkt: ∼ 275 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006753
CAS-Nummer: 345909-35-5
EINECS: 207-751-5
Merck Index: 14,5327

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR