2-Phenyl-2-oxazolin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Phenyl(2-oxazoline), (4,5-Dihydro-2-oxazolyl)benzene, 4,5-Dihydro-2-phenyloxazole
M-Clarity™ Qualitätsstufe = MQ100
369756-100G
369756-25G
SIAL369756-100GEA
381
EUR
SIAL369756-100G
SIAL369756-25G
2-Phenyl-2-oxazolin, Sigma-Aldrich®
2-Phenyl-2-oxazolin
Formel:
C₉H₉NO Molecular Weight: 147,1739 g/mol |
MDL:
MFCD00075541 CAS-Nummer: 7127-19-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...