Menadion (2-Methyl-1,4-naphthochinon, Vitamin K3) ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Menadione, 1,4-Dihydro-2-methylnaphthalene-1,4-dione, 2-Methylnaphthalene-1,4-dione

A13593.09 A13593.18 A13593.30
A13593.09EA 24.8 EUR
A13593.09 A13593.18 A13593.30
Menadion (2-Methyl-1,4-naphthochinon, Vitamin K3) ≥98%
Menadion (2-Methyl-1,4-naphthochinon, Vitamin K3)

Purity: 98% CAS No.: 58-27-5 Molecular Formula: C11H8O2 Formula Weight: 172.18 Odour: Odourless Melting Point: 104-107° Beilstein Reference: 1908453 Merck Reference: 14,5831 MDL No.: MFCD00001681

Formel: C₁₁H₈O₂
Molecular Weight: 172,19 g/mol
Schmelzpunkt: 104…107 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001681
CAS-Nummer: 58-27-5
EINECS: 200-372-6
Merck Index: 14,5831

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR