Menadion (2-Methyl-1,4-naphthochinon, Vitamin K3), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Menadione, 1,4-Dihydro-2-methylnaphthalene-1,4-dione, 2-Methylnaphthalene-1,4-dione

M-Clarity™ Qualitätsstufe = MQ200

M9429-100G M9429-25G
SIALM9429-100GEA 287 EUR
SIALM9429-100G SIALM9429-25G
Menadion (2-Methyl-1,4-naphthochinon, Vitamin K3), Sigma-Aldrich®
Menadion (2-Methyl-1,4-naphthochinon, Vitamin K3)
Formel: C₁₁H₈O₂
Molecular Weight: 172.19 g/mol
MDL: MFCD00001681
CAS-Nummer: 58-27-5
EINECS: 200-372-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR