α-Naphtholphthalein
Lieferant: Merck
Synonyme:
3,3-Bis(4-hydroxynaphthalen-1-yl)isobenzofuran-1(3H)-one
M-Clarity™ Qualitätsstufe = MQ200
1.06246.0001
1.06246.0005
1.06246.0001EA
328
EUR
1.06246.0001
1.06246.0005
α-Naphtholphthalein
α-Naphtholphthalein
Formel:
C₂₈H₁₈O₄ Molecular Weight: 418,45 g/mol |
MDL:
MFCD00036202 CAS-Nummer: 596-01-0 EINECS: 209-875-5 |
Spezifikation Testergebnisse
Solubility | (20 °C) practically insoluble |
Melting Point | 253 - 255 °C |
Molar Mass | 418.45 g/mol |
Bulk Density | 350 kg/m3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...