2-(3-Bromphenyl)-5-tert-butyl-1,3,4-oxadiazol

Lieferant: COMBI-BLOCKS

HC-6399-1G HC-6399-5G
COBBHC-6399-1GEA 149 EUR
COBBHC-6399-1G COBBHC-6399-5G
2-(3-Bromphenyl)-5-tert-butyl-1,3,4-oxadiazol
2-(3-Bromphenyl)-5-tert-butyl-1,3,4-oxadiazol
Formel: C₁₂H₁₃BrN₂O
MDL: MFCD09878391
CAS-Nummer: 957065-96-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR