2-(3-Bromphenyl)-5-butyl-1,3,4-oxadiazol

Lieferant: COMBI-BLOCKS

HC-6400-5G HC-6400-1G
COBBHC-6400-5GEA 451 EUR
COBBHC-6400-5G COBBHC-6400-1G
2-(3-Bromphenyl)-5-butyl-1,3,4-oxadiazol
2-(3-Bromophenyl)-5-butyl-1,3,4-oxadiazole
Formel: C₁₂H₁₃BrN₂O
MDL: MFCD09878392
CAS-Nummer: 957065-94-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR