2-(3-Bromphenyl)-5-ethyl-1,3,4-oxadiazol

Lieferant: COMBI-BLOCKS

HC-6377-5G HC-6377-1G
COBBHC-6377-5GEA 451 EUR
COBBHC-6377-5G COBBHC-6377-1G
2-(3-Bromphenyl)-5-ethyl-1,3,4-oxadiazol
2-(3-Bromophenyl)-5-ethyl-1,3,4-oxadiazole
Formel: C₁₀H₉BrN₂O
MDL: MFCD09878384
CAS-Nummer: 957065-88-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR